Product Details

SNP ID
rs748651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1017327 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCCAGCTCCTCCATGCGGGAACT[C/T]GGGTAGCCGGTCAGGGTTGGGGTTG
Phenotype
MIM: 605830
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGFRL1 PubMed Links

Gene Details

Gene
FGFRL1
Gene Name
fibroblast growth factor receptor-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004356.2 Intron NP_001004356.1
NM_001004358.1 Intron NP_001004358.1
NM_021923.3 Intron NP_068742.2
XM_011513486.1 Intron XP_011511788.1

View Full Product Details