Product Details

SNP ID
rs1564508
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:1251091 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCGTCACTCTCCATGGCCTCCGTCT[A/G]CACACAGTGTTCGTCTGCACAGCTT
Phenotype
MIM: 602618
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTBP1 PubMed Links

Gene Details

Gene
CTBP1
Gene Name
C-terminal binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012614.1 Intron NP_001012632.1
NM_001328.2 Intron NP_001319.1
XM_005272261.3 Intron XP_005272318.1
XM_005272263.4 Intron XP_005272320.1
XM_011513395.2 Intron XP_011511697.1
XM_011513396.2 Intron XP_011511698.1
XM_011513397.2 Intron XP_011511699.1
XM_017007762.1 Intron XP_016863251.1
XM_017007763.1 Intron XP_016863252.1
XM_017007764.1 Intron XP_016863253.1
XM_017007765.1 Intron XP_016863254.1
XM_017007766.1 Intron XP_016863255.1
XM_017007767.1 Intron XP_016863256.1
Gene
CTBP1-AS2
Gene Name
CTBP1 antisense RNA 2 (head to head)
There are no transcripts associated with this gene.

View Full Product Details