Product Details

SNP ID
rs1188817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87346001 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATAACCTTTAAATAGTAGCTGGT[A/G]GCATAAAAGTAGATACATATAGATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf163 PubMed Links

Gene Details

Gene
C6orf163
Gene Name
chromosome 6 open reading frame 163
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010868.2 96 Intron NP_001010868.2
XM_005248673.3 96 Intron XP_005248730.1
XM_006715370.3 96 Intron XP_006715433.1
XM_006715372.2 96 Intron XP_006715435.1
XM_011535539.1 96 Intron XP_011533841.1
XM_017010367.1 96 UTR 5 XP_016865856.1
Gene
SMIM8
Gene Name
small integral membrane protein 8
There are no transcripts associated with this gene.

View Full Product Details