Product Details

SNP ID
rs7984311
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:100089498 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGGTGCCCGCACGCCCTCCGAGGC[A/G]GCTGATGGCTTTGGGCAGGCATATT
Phenotype
MIM: 232000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105370333 PubMed Links

Gene Details

Gene
LOC105370333
Gene Name
uncharacterized LOC105370333
There are no transcripts associated with this gene.

Gene
PCCA
Gene Name
propionyl-CoA carboxylase alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000282.3 Intron NP_000273.2
NM_001127692.2 Intron NP_001121164.1
NM_001178004.1 Intron NP_001171475.1
XM_005254059.2 Intron XP_005254116.1
XM_011521093.1 Intron XP_011519395.1
XM_017020605.1 Intron XP_016876094.1
XM_017020606.1 Intron XP_016876095.1
XM_017020607.1 Intron XP_016876096.1
XM_017020608.1 Intron XP_016876097.1
XM_017020609.1 Intron XP_016876098.1
XM_017020610.1 Intron XP_016876099.1
XM_017020611.1 Intron XP_016876100.1
XM_017020612.1 Intron XP_016876101.1
XM_017020613.1 Intron XP_016876102.1
XM_017020614.1 Intron XP_016876103.1
XM_017020615.1 Intron XP_016876104.1
XM_017020616.1 Intron XP_016876105.1
XM_017020617.1 Intron XP_016876106.1
XM_017020618.1 Intron XP_016876107.1

View Full Product Details