Product Details

SNP ID
rs1054772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18705436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTAGACCAGTTTTAATTTTTATT[G/T]CACATCAGAATCACCCGCAAAATTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IGSF22 PubMed Links

Gene Details

Gene
IGSF22
Gene Name
immunoglobulin superfamily member 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173588.3 Intron NP_775859.3
XM_011520030.2 Intron XP_011518332.1
XM_011520031.2 Intron XP_011518333.1
XM_017017607.1 Intron XP_016873096.1
XM_017017608.1 Intron XP_016873097.1
Gene
TMEM86A
Gene Name
transmembrane protein 86A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153347.2 Intron NP_699178.1

View Full Product Details