Product Details
- SNP ID
-
rs8039661
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:78966667 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTAAAAAAGCCAAGTAGCATTAAA[C/T]GGTTCATAATGAAAACCAGCAATCC
- Phenotype
-
MIM: 606600
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RASGRF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16970371] are located under a probe and SNP(s) [rs79231687] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RASGRF1
- Gene Name
- Ras protein specific guanine nucleotide releasing factor 1
View Full Product Details