Product Details
- SNP ID
-
rs7613431
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:178539801 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGTGATTTCTCCTTATTTTATAAA[C/T]AGAGACTTTTTCAATCTCCTGCTGG
- Phenotype
-
MIM: 605214
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KCNMB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115497706] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCNMB2
- Gene Name
- potassium calcium-activated channel subfamily M regulatory beta subunit 2
- Gene
- KCNMB2-AS1
- Gene Name
- KCNMB2 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details