Product Details

SNP ID
rs7065102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13731661 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCTACAACAGTGCCAGGGACCCA[A/G]AAAGCACTCATTATTTGCTGAATGA
Phenotype
MIM: 300170 MIM: 300202
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OFD1 PubMed Links

Gene Details

Gene
OFD1
Gene Name
OFD1, centriole and centriolar satellite protein
There are no transcripts associated with this gene.

Gene
TRAPPC2
Gene Name
trafficking protein particle complex 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011658.3 Intron NP_001011658.1
NM_001128835.2 Intron NP_001122307.2
NM_014563.5 Intron NP_055378.1
XM_011545565.1 Intron XP_011543867.1
XM_011545566.2 Intron XP_011543868.1

View Full Product Details