Product Details

SNP ID
rs12713802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73998137 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTGAGAATCCTAGAGGTGGAAGGT[C/T]AGACGCTTTGTGTTAATTCCTCTTG
Phenotype
MIM: 613555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TET3 PubMed Links

Gene Details

Gene
TET3
Gene Name
tet methylcytosine dioxygenase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287491.1 Intron NP_001274420.1
XM_005264187.3 Intron XP_005264244.1
XM_011532682.2 Intron XP_011530984.1
XM_011532683.2 Intron XP_011530985.1
XM_011532684.2 Intron XP_011530986.1
XM_011532685.2 Intron XP_011530987.1
XM_011532686.2 Intron XP_011530988.1
XM_011532687.2 Intron XP_011530989.1
XM_011532688.2 Intron XP_011530990.1
XM_011532689.2 Intron XP_011530991.1
XM_011532690.1 Intron XP_011530992.1
XM_017003566.1 Intron XP_016859055.1
XM_017003567.1 Intron XP_016859056.1

View Full Product Details