Product Details

SNP ID
rs12832688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:82359583 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTGAGAGGCCAGTTAGGAGACTA[A/C]CTCAGTAGTACAACAGATGATTGTA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC59 PubMed Links

Gene Details

Gene
CCDC59
Gene Name
coiled-coil domain containing 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014167.4 Intron NP_054886.2
Gene
METTL25
Gene Name
methyltransferase like 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319675.1 Intron NP_001306604.1
NM_032230.2 Intron NP_115606.2
XM_005269186.4 Intron XP_005269243.2
XM_011538827.2 Intron XP_011537129.1
XM_011538828.2 Intron XP_011537130.1
XM_011538829.2 Intron XP_011537131.1
XM_017020020.1 Intron XP_016875509.1
XM_017020021.1 Intron XP_016875510.1

View Full Product Details