Product Details

SNP ID
rs10870159
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:136992590 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GAGTGGGGGTCCGCGGGGAGGTAGG[C/G]GTGCAGGGAGCTCCGCGGGCGGCCC
Phenotype
MIM: 616315 MIM: 606533
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C9orf142 PubMed Links

Gene Details

Gene
C9orf142
Gene Name
chromosome 9 open reading frame 142
There are no transcripts associated with this gene.

Gene
CLIC3
Gene Name
chloride intracellular channel 3
There are no transcripts associated with this gene.

Gene
LCNL1
Gene Name
lipocalin like 1
There are no transcripts associated with this gene.

View Full Product Details