Product Details

SNP ID
rs34418989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123537075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCTTCTCAATTCCAAAAGTGAA[A/T]GAGTAATTTTACAATACCATTTTCC
Phenotype
MIM: 601677
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IQUB PubMed Links

Gene Details

Gene
IQUB
Gene Name
IQ motif and ubiquitin domain containing
There are no transcripts associated with this gene.

Gene
NDUFA5
Gene Name
NADH:ubiquinone oxidoreductase subunit A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282419.2 5698 UTR 3 NP_001269348.1
NM_001282420.2 5698 UTR 3 NP_001269349.1
NM_001282421.2 5698 UTR 3 NP_001269350.1
NM_001282422.2 5698 UTR 3 NP_001269351.1
NM_001291304.1 5698 UTR 3 NP_001278233.1
NM_005000.4 5698 UTR 3 NP_004991.1

View Full Product Details