Product Details

SNP ID
rs4955418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49163194 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGCTTTAGCCTTGGCCTTGGCCC[A/G]TGCTGCCATGACTTTGGCCTTGGCC
Phenotype
MIM: 613169
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC71 PubMed Links

Gene Details

Gene
CCDC71
Gene Name
coiled-coil domain containing 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022903.3 1153 Missense Mutation NP_075054.3
Gene
KLHDC8B
Gene Name
kelch domain containing 8B
There are no transcripts associated with this gene.

Gene
LAMB2P1
Gene Name
laminin subunit beta 2 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details