Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001271049.1 | 218 | Intron | NP_001257978.1 | ||
XM_006712353.3 | 218 | Intron | XP_006712416.1 | ||
XM_017003550.1 | 218 | Intron | XP_016859039.1 | ||
XM_017003551.1 | 218 | Intron | XP_016859040.1 | ||
XM_017003552.1 | 218 | Intron | XP_016859041.1 | ||
XM_017003553.1 | 218 | Intron | XP_016859042.1 | ||
XM_017003554.1 | 218 | Intron | XP_016859043.1 | ||
XM_017003555.1 | 218 | Intron | XP_016859044.1 | ||
XM_017003556.1 | 218 | Intron | XP_016859045.1 | ||
XM_017003557.1 | 218 | Intron | XP_016859046.1 | ||
XM_017003558.1 | 218 | Intron | XP_016859047.1 | ||
XM_017003559.1 | 218 | Intron | XP_016859048.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001105198.1 | 218 | Missense Mutation | GCA,GGA | A29G | NP_001098668.1 |
NM_001105199.1 | 218 | Missense Mutation | GCA,GGA | A29G | NP_001098669.1 |
NM_030577.2 | 218 | Missense Mutation | GCA,GGA | A29G | NP_085054.2 |