Product Details

SNP ID
rs4076819
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:81559543 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCCCAACATCCCACCCAGTGCTGGT[C/G]CTGCCCACACAGCACTCCCACCACA
Phenotype
MIM: 611301 MIM: 606590
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAAP100 PubMed Links

Gene Details

Gene
FAAP100
Gene Name
Fanconi anemia core complex associated protein 100
There are no transcripts associated with this gene.

Gene
NPLOC4
Gene Name
NPL4 homolog, ubiquitin recognition factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017921.3 Intron NP_060391.2
XM_011524979.1 Intron XP_011523281.1
XM_011524980.1 Intron XP_011523282.1
XM_011524981.1 Intron XP_011523283.1
XM_011524982.2 Intron XP_011523284.1
XM_017024830.1 Intron XP_016880319.1

View Full Product Details