Product Details

SNP ID
rs4460634
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:159872901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGCCTGTGGTGCCATCTCTTTG[C/G]GGGAGGGAAGAGGATTTGGGAAAGG
Phenotype
MIM: 605152
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFAP45 PubMed Links

Gene Details

Gene
CFAP45
Gene Name
cilia and flagella associated protein 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012337.2 Intron NP_036469.2

View Full Product Details