Product Details
- SNP ID
-
rs4077991
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:8275019 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ATCTGGGACCACTCAGGCAAGATGA[A/C]GCCTCCCCTCCTGCCCCGCCCACTC
- Phenotype
-
MIM: 608785
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HTRA3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116746566] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HTRA3
- Gene Name
- HtrA serine peptidase 3
View Full Product Details