Product Details

SNP ID
rs3735086
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:150626559 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCAGTATCAGTACTTATAAACCCC[A/G]TCTGTAGGAACAGCCTTCCAGGAAA
Phenotype
MIM: 616960
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GIMAP6 PubMed Links

Gene Details

Gene
GIMAP6
Gene Name
GTPase, IMAP family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244071.1 2295 UTR 3 NP_001231000.1
NM_001244072.1 2295 UTR 3 NP_001231001.1
NM_024711.5 2295 UTR 3 NP_078987.3

View Full Product Details