Product Details

SNP ID
rs12706486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123118136 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGCAACAGAGCTCAGGTAGGTTT[G/T]CAAGTCCCTGTGGCTCCAGCAGTTC
Phenotype
MIM: 606193
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC13A1 PubMed Links

Gene Details

Gene
SLC13A1
Gene Name
solute carrier family 13 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324400.1 Intron NP_001311329.1
NM_022444.3 Intron NP_071889.2
XM_011516515.2 Intron XP_011514817.1
XM_011516516.2 Intron XP_011514818.1
XM_011516517.2 Intron XP_011514819.1
XM_011516518.2 Intron XP_011514820.1
XM_011516519.2 Intron XP_011514821.1
XM_017012554.1 Intron XP_016868043.1
XM_017012555.1 Intron XP_016868044.1

View Full Product Details