Product Details

SNP ID
rs13254148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142199381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAAACCCCCTGGGCTGCCCACA[C/T]TTCCATGGGGTCACCTGCCCTCTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00051 PubMed Links

Gene Details

Gene
LINC00051
Gene Name
long intergenic non-protein coding RNA 51
There are no transcripts associated with this gene.

View Full Product Details