Product Details
- SNP ID
-
rs9660679
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
40
- Location
-
Chr.1:153609782 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CCCTGATAACGGGTATTGCACGTCC[C/T]TGACCACCTTTTAAGCTTTTTCTTT
- Phenotype
-
MIM: 601989
MIM: 607986
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
S100A13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9660416] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- S100A13
- Gene Name
- S100 calcium binding protein A13
There are no transcripts associated with this gene.
- Gene
- S100A14
- Gene Name
- S100 calcium binding protein A14
There are no transcripts associated with this gene.
- Gene
- S100A16
- Gene Name
- S100 calcium binding protein A16
View Full Product Details