Product Details

SNP ID
rs10838011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1630791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGATCCAAAGCGGCCCACTGAGG[A/C]CCCAGAGGCAGATCAGACCCCTTAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP5-5 PubMed Links

Gene Details

Gene
KRTAP5-5
Gene Name
keratin associated protein 5-5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001480.2 989 UTR 3 NP_001001480.2

View Full Product Details