Product Details

SNP ID
rs6439021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126664517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGACCATTTCCACCTTCTGAC[A/G]AAGGCTCAGGGCTAAAATTGGGGCC
Phenotype
MIM: 606235
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUP210P1 PubMed Links

Gene Details

Gene
NUP210P1
Gene Name
nucleoporin 210 pseudogene 1
There are no transcripts associated with this gene.

Gene
TXNRD3
Gene Name
thioredoxin reductase 3
There are no transcripts associated with this gene.

View Full Product Details