Product Details

SNP ID
rs11994518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143214242 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAGTGGGGGCCTAGGGTGGAGAC[A/G]CAAGCACAGAGGCCCCAGGGTAGGG
Phenotype
MIM: 612757
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPIHBP1 PubMed Links

Gene Details

Gene
GPIHBP1
Gene Name
glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301772.1 Intron NP_001288701.1
NM_178172.5 Intron NP_835466.2

View Full Product Details