Product Details

SNP ID
rs9302325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:38258998 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAATTTGGACTAAGCATCATATG[C/T]GCCCTTCCTTTGTTGATGGTGTATG
Phenotype
MIM: 609291
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPRED1 PubMed Links

Gene Details

Gene
SPRED1
Gene Name
sprouty related EVH1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152594.2 Intron NP_689807.1
XM_005254202.3 Intron XP_005254259.1
XM_011521289.2 Intron XP_011519591.1

View Full Product Details