Product Details

SNP ID
rs3184121
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:124967889 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGAGGAAAAGACAATGGCAATAAGA[A/C]CCTCTGGAGCTTAAACCAAACCACT
Phenotype
MIM: 614182
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HEG1 PubMed Links

Gene Details

Gene
HEG1
Gene Name
heart development protein with EGF like domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020733.1 7272 UTR 3 NP_065784.1
XM_005247666.1 7272 UTR 3 XP_005247723.1

View Full Product Details