Product Details

SNP ID
rs1526505
Assay Type
Validated
NCBI dbSNP Submissions
45
Location
Chr.1:75133215 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAAAAACAATGGCGATAGGAAAATG[C/T]TCTGAAGACTCTAGTGTTTATGGGA
Phenotype
MIM: 604425
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LHX8 PubMed Links

Gene Details

Gene
LHX8
Gene Name
LIM homeobox 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001933.1 Intron NP_001001933.1
NM_001256114.1 Intron NP_001243043.1
XM_017001316.1 Intron XP_016856805.1
XM_017001317.1 Intron XP_016856806.1

View Full Product Details