Product Details

SNP ID
rs13406651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:166901062 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGCCTTTGCTGATGGGAGTAAGG[A/G]TTTAGTTGTGGTTTTTCATGTTGTT
Phenotype
MIM: 609778
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XIRP2 PubMed Links

Gene Details

Gene
XIRP2
Gene Name
xin actin binding repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079810.3 Intron NP_001073278.1
NM_001199143.1 Intron NP_001186072.1
NM_001199144.1 Intron NP_001186073.1
NM_001199145.1 Intron NP_001186074.1
NM_152381.5 Intron NP_689594.4
XM_017003309.1 Intron XP_016858798.1

View Full Product Details