Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001302826.1 | 985 | Silent Mutation | CTA,CTG | L254L | NP_001289755.1 |
NM_001302827.1 | 985 | UTR 3 | NP_001289756.1 | ||
NM_016229.4 | 985 | Silent Mutation | CTA,CTG | L254L | NP_057313.2 |
XM_005252975.4 | 985 | Silent Mutation | CTA,CTG | L254L | XP_005253032.1 |
XM_006718251.2 | 985 | Silent Mutation | CTA,CTG | L254L | XP_006718314.1 |
XM_011520184.2 | 985 | Silent Mutation | CTA,CTG | L314L | XP_011518486.1 |
XM_011520185.2 | 985 | UTR 3 | XP_011518487.1 | ||
XM_017017920.1 | 985 | Silent Mutation | CTA,CTG | L314L | XP_016873409.1 |
XM_017017921.1 | 985 | UTR 3 | XP_016873410.1 | ||
XM_017017922.1 | 985 | UTR 3 | XP_016873411.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001256568.1 | 985 | Intron | NP_001243497.1 | ||
NM_001256569.1 | 985 | Intron | NP_001243498.1 | ||
NM_003621.3 | 985 | Intron | NP_003612.2 | ||
XM_005253177.2 | 985 | Intron | XP_005253234.1 | ||
XM_006718349.2 | 985 | Intron | XP_006718412.1 | ||
XM_006718350.2 | 985 | Intron | XP_006718413.1 | ||
XM_006718351.2 | 985 | Intron | XP_006718414.1 | ||
XM_006718352.2 | 985 | Intron | XP_006718415.1 | ||
XM_006718353.2 | 985 | Intron | XP_006718416.1 | ||
XM_011520413.2 | 985 | Intron | XP_011518715.1 | ||
XM_011520414.2 | 985 | Intron | XP_011518716.1 | ||
XM_011520415.2 | 985 | Intron | XP_011518717.1 | ||
XM_011520416.2 | 985 | Intron | XP_011518718.1 | ||
XM_011520417.2 | 985 | Intron | XP_011518719.1 | ||
XM_011520418.2 | 985 | Intron | XP_011518720.1 | ||
XM_011520419.2 | 985 | Intron | XP_011518721.1 | ||
XM_011520420.2 | 985 | Intron | XP_011518722.1 | ||
XM_017018443.1 | 985 | Intron | XP_016873932.1 | ||
XM_017018444.1 | 985 | Intron | XP_016873933.1 | ||
XM_017018445.1 | 985 | Intron | XP_016873934.1 | ||
XM_017018446.1 | 985 | Intron | XP_016873935.1 | ||
XM_017018447.1 | 985 | Intron | XP_016873936.1 |