Product Details

SNP ID
rs13073206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:130220778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTTCATGGCTCTGTGGGCGCTA[C/T]GACACAGGAATGGCACAGATCATGG
Phenotype
MIM: 616612
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL6A4P2 PubMed Links

Gene Details

Gene
COL6A4P2
Gene Name
collagen type VI alpha 4 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details