Product Details

SNP ID
rs6576108
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:105493530 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCCTGTCCAGAGGCTGTTCCTCCCC[C/T]GATGCTGCTGGGGAAGCCAGGGCAC
Phenotype
MIM: 123875
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf80 PubMed Links

Gene Details

Gene
C14orf80
Gene Name
chromosome 14 open reading frame 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134875.1 Intron NP_001128347.1
NM_001134876.1 Intron NP_001128348.1
NM_001134877.1 Intron NP_001128349.1
NM_001198983.1 Intron NP_001185912.1
Gene
CRIP1
Gene Name
cysteine rich protein 1
There are no transcripts associated with this gene.

View Full Product Details