Product Details

SNP ID
rs1042752
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:111352386 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTACCAACTGAAATGTCTGCTTAAT[A/G]GTGACGATGGTGCTATTAGCAGGTG
Phenotype
MIM: 601206
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4491 PubMed Links

Gene Details

Gene
MIR4491
Gene Name
microRNA 4491
There are no transcripts associated with this gene.

Gene
POU2AF1
Gene Name
POU class 2 associating factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006235.2 4602 UTR 3 NP_006226.2
XM_005271593.1 4602 UTR 3 XP_005271650.1
XM_005271594.3 4602 UTR 3 XP_005271651.1
XM_006718859.1 4602 UTR 3 XP_006718922.1
XM_006718860.3 4602 UTR 3 XP_006718923.1
XM_017017932.1 4602 UTR 3 XP_016873421.1

View Full Product Details