Product Details

SNP ID
rs11900255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:138671865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGAAAACTGCGAGTATGAAAAT[G/T]AAAAATGGTGAGGTTAAGAACAAAG
Phenotype
MIM: 604635
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NXPH2 PubMed Links

Gene Details

Gene
NXPH2
Gene Name
neurexophilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007226.2 Intron NP_009157.1

View Full Product Details