Product Details

SNP ID
rs11121529
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:10211630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGGGGTTGGGAGGCTGCTTGAG[C/G]AAGCTAGTCATTGTTCATGCAACAC
Phenotype
MIM: 605995
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIF1B PubMed Links

Gene Details

Gene
KIF1B
Gene Name
kinesin family member 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015074.3 Intron NP_055889.2
NM_183416.3 Intron NP_904325.2

View Full Product Details