Product Details

SNP ID
rs34090087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:199771489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATAGTTCTTTGAACAAGAATT[A/C]ATGATAAATACACATGTGTAAAAGA
Phenotype
MIM: 614308
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FTCDNL1 PubMed Links

Gene Details

Gene
FTCDNL1
Gene Name
formiminotransferase cyclodeaminase N-terminal like
There are no transcripts associated with this gene.

View Full Product Details