Product Details

SNP ID
rs28483505
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:58766748 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TATGAGATGACAATCATTAAATTCA[C/G]CTATCACTGATTTTGCAAAAGGAAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PPM1E PubMed Links

Gene Details

Gene
PPM1E
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014906.4 Intron NP_055721.3
XM_005257169.4 Intron XP_005257226.1
XM_011524534.2 Intron XP_011522836.1
XM_017024359.1 Intron XP_016879848.1

View Full Product Details