Product Details

SNP ID
rs13109244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:176214998 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGTAACAAGTCCATGTACTATGG[A/G]GAAGAGAAGTGACTTGTGAGTCCTT
Phenotype
MIM: 615050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB5 PubMed Links

Gene Details

Gene
ASB5
Gene Name
ankyrin repeat and SOCS box containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080874.3 2397 UTR 3 NP_543150.1
XM_005262759.1 2397 UTR 3 XP_005262816.1
XM_011531616.2 2397 UTR 3 XP_011529918.1
XM_011531617.2 2397 UTR 3 XP_011529919.1

View Full Product Details