Product Details

SNP ID
rs1574340
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:125083833 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGAAGCAGCTCCACGAAGGTCCTG[A/G]GCTTGGGTCCACTGTACATGGGACA
Phenotype
MIM: 611316
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC12A8 PubMed Links

Gene Details

Gene
SLC12A8
Gene Name
solute carrier family 12 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195483.1 2252 UTR 3 NP_001182412.1
NM_024628.5 2252 UTR 3 NP_078904.3

View Full Product Details