Product Details
- SNP ID
-
rs7323023
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:38968394 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCCCTCCATGTGTGAACTGCACAAC[A/C]CTTACCTTGGCTCTCGCTTCCCGGG
- Phenotype
-
MIM: 608327
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
STOML3
PubMed Links
Gene Details
- Gene
- STOML3
- Gene Name
- stomatin like 3
View Full Product Details