Product Details
- SNP ID
-
rs2272501
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:81262082 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGGCTTTAGAGGGACTTGGATGATT[C/G]AGTACAAGATTTCAGATGAGAAAGA
- Phenotype
-
MIM: 614356
MIM: 603143
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ACSS3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10862276] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACSS3
- Gene Name
- acyl-CoA synthetase short-chain family member 3
There are no transcripts associated with this gene.
- Gene
- PPFIA2
- Gene Name
- PTPRF interacting protein alpha 2
View Full Product Details