Product Details

SNP ID
rs6707397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112141932 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTTCTGGGTCCAATCCCTGTCCC[T/G]CCACATCCAGTCTTTCTCAAGTTTC
Phenotype
MIM: 611551
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
FBLN7 PubMed Links

Gene Details

Gene
FBLN7
Gene Name
fibulin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128165.1 Intron NP_001121637.1
NM_153214.2 Intron NP_694946.2
XM_006712260.3 Intron XP_006712323.1
XM_011510585.2 Intron XP_011508887.1
XM_011510587.2 Intron XP_011508889.1
XM_017003317.1 Intron XP_016858806.1
XM_017003318.1 Intron XP_016858807.1
XM_017003319.1 Intron XP_016858808.1

View Full Product Details