Product Details

SNP ID
rs34061491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111902319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAGAAGACCCTGATTGCCCGTA[A/G]GTTAAGAGGTCAGCATTGAGGTCAC
Phenotype
MIM: 606723
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM1A PubMed Links

Gene Details

Gene
ADAM1A
Gene Name
ADAM metallopeptidase domain 1A (pseudogene)
There are no transcripts associated with this gene.

Gene
MAPKAPK5
Gene Name
mitogen-activated protein kinase-activated protein kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003668.3 Intron NP_003659.2
NM_139078.2 Intron NP_620777.1
XM_017020125.1 Intron XP_016875614.1
XM_017020126.1 Intron XP_016875615.1
XM_017020127.1 Intron XP_016875616.1
XM_017020128.1 Intron XP_016875617.1
XM_017020129.1 Intron XP_016875618.1
XM_017020130.1 Intron XP_016875619.1
XM_017020131.1 Intron XP_016875620.1
XM_017020132.1 Intron XP_016875621.1
XM_017020133.1 Intron XP_016875622.1
XM_017020134.1 Intron XP_016875623.1
XM_017020135.1 Intron XP_016875624.1
XM_017020136.1 Intron XP_016875625.1
XM_017020137.1 Intron XP_016875626.1
XM_017020138.1 Intron XP_016875627.1
XM_017020139.1 Intron XP_016875628.1
Gene
TMEM116
Gene Name
transmembrane protein 116
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193453.1 Intron NP_001180382.1
NM_001193531.1 Intron NP_001180460.1
NM_001294314.1 Intron NP_001281243.1
NM_138341.2 Intron NP_612350.1
XM_006719678.3 Intron XP_006719741.1
XM_011538946.1 Intron XP_011537248.1
XM_011538947.1 Intron XP_011537249.1
XM_011538948.2 Intron XP_011537250.1
XM_011538949.2 Intron XP_011537251.1
XM_011538950.1 Intron XP_011537252.1
XM_011538951.2 Intron XP_011537253.1
XM_011538953.1 Intron XP_011537255.1
XM_011538955.1 Intron XP_011537257.1
XM_011538957.1 Intron XP_011537259.1
XM_011538958.1 Intron XP_011537260.1
XM_017020187.1 Intron XP_016875676.1
XM_017020188.1 Intron XP_016875677.1
XM_017020189.1 Intron XP_016875678.1
XM_017020190.1 Intron XP_016875679.1
XM_017020191.1 Intron XP_016875680.1
XM_017020192.1 Intron XP_016875681.1
XM_017020193.1 Intron XP_016875682.1
XM_017020194.1 Intron XP_016875683.1

View Full Product Details