Product Details

SNP ID
rs5764372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43882872 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGACCTTCTTCCAGAGAATTTTT[A/G]AAAATACAGGTCCCCCCATGACACC
Phenotype
MIM: 611589
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PNPLA5 PubMed Links

Gene Details

Gene
PNPLA5
Gene Name
patatin like phospholipase domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177675.1 Intron NP_001171146.1
NM_138814.3 Intron NP_620169.1
XM_011529959.2 Intron XP_011528261.1

View Full Product Details