Product Details

SNP ID
rs2239914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35632501 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTGCCTAACACTTCATTGCCCCT[C/T]GGGTCTCAGCTCACAGGTCCCTTTC
Phenotype
MIM: 609358 MIM: 613432 MIM: 613232
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ETV2 PubMed Links

Gene Details

Gene
ETV2
Gene Name
ETS variant 2
There are no transcripts associated with this gene.

Gene
HAUS5
Gene Name
HAUS augmin like complex subunit 5
There are no transcripts associated with this gene.

Gene
RBM42
Gene Name
RNA binding motif protein 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319113.1 Intron NP_001306042.1
NM_024321.4 Intron NP_077297.2
XM_005259257.2 Intron XP_005259314.1

View Full Product Details