Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001280542.1 | 6428 | Intron | NP_001267471.1 | ||
NM_001280543.1 | 6428 | Intron | NP_001267472.1 | ||
NM_001280544.1 | 6428 | Intron | NP_001267473.1 | ||
NM_012074.4 | 6428 | Intron | NP_036206.3 | ||
XM_017021670.1 | 6428 | UTR 3 | XP_016877159.1 | ||
XM_017021671.1 | 6428 | Intron | XP_016877160.1 | ||
XM_017021672.1 | 6428 | Intron | XP_016877161.1 | ||
XM_017021673.1 | 6428 | Intron | XP_016877162.1 | ||
XM_017021674.1 | 6428 | Intron | XP_016877163.1 | ||
XM_017021675.1 | 6428 | Intron | XP_016877164.1 |