Product Details

SNP ID
rs2299014
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:132567707 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GAGTGTTGTATTCTAAAGGTCACAC[A/C]ATGCTGTGAGACATTTGAGTCCTGA
Phenotype
MIM: 604040
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RAD50 PubMed Links

Gene Details

Gene
RAD50
Gene Name
RAD50 double strand break repair protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005732.3 Intron NP_005723.2

View Full Product Details