Product Details

SNP ID
rs12472035
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202221970 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGTCCTCTGTAACTGGAAGGAGA[A/G]GAGGCCTTAAGAAGCTTTGTTTAAG
Phenotype
MIM: 601912
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SUMO1 PubMed Links

Gene Details

Gene
SUMO1
Gene Name
small ubiquitin-like modifier 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005781.1 Intron NP_001005781.1
NM_001005782.1 Intron NP_001005782.1
NM_003352.4 Intron NP_003343.1

View Full Product Details