Product Details

SNP ID
rs2713603
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:128481691 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CCCTCTTACGGGAAGCCCTTCTGGC[A/G]CTCACTCAGGGCAGCAGCTTCCCAA
Phenotype
MIM: 137295
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB8-AS1 PubMed Links

Gene Details

Gene
DNAJB8-AS1
Gene Name
DNAJB8 antisense RNA 1
There are no transcripts associated with this gene.

Gene
GATA2
Gene Name
GATA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145661.1 Intron NP_001139133.1
NM_001145662.1 Intron NP_001139134.1
NM_032638.4 Intron NP_116027.2
Gene
GATA2-AS1
Gene Name
GATA2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details