Product Details

SNP ID
rs35753959
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:200406635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATTCCCATGCGTTATTACACT[G/T]TCCTGAGAGTAAAGCAATTAGAATA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF281 PubMed Links

Gene Details

Gene
ZNF281
Gene Name
zinc finger protein 281
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281293.1 4583 UTR 3 NP_001268222.1
NM_001281294.1 4583 UTR 3 NP_001268223.1
NM_012482.4 4583 UTR 3 NP_036614.1
XM_017000887.1 4583 UTR 3 XP_016856376.1
XM_017000888.1 4583 UTR 3 XP_016856377.1

View Full Product Details