Product Details

SNP ID
rs12414205
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:32451112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGCAGGAAATGCTATAAGTCCT[A/G]TAGGTAAAATAATCCAACATTTTCC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CCDC7 PubMed Links

Gene Details

Gene
CCDC7
Gene Name
coiled-coil domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001026383.2 Intron NP_001021554.1
NM_001321115.1 Intron NP_001308044.1
NM_145023.5 Intron NP_659460.3
XM_005252593.2 Intron XP_005252650.1
XM_006717505.1 Intron XP_006717568.1
XM_006717506.1 Intron XP_006717569.1
XM_011519663.1 Intron XP_011517965.1
XM_011519664.1 Intron XP_011517966.1
XM_011519666.1 Intron XP_011517968.1
XM_011519667.1 Intron XP_011517969.1
XM_011519668.1 Intron XP_011517970.1
XM_011519669.1 Intron XP_011517971.1
XM_011519670.1 Intron XP_011517972.1
XM_011519671.1 Intron XP_011517973.1
XM_011519672.1 Intron XP_011517974.1
XM_011519673.1 Intron XP_011517975.1
XM_011519674.1 Intron XP_011517976.1
XM_011519675.1 Intron XP_011517977.1
XM_011519676.1 Intron XP_011517978.1
XM_011519677.1 Intron XP_011517979.1
XM_011519678.1 Intron XP_011517980.1
XM_011519679.1 Intron XP_011517981.1
XM_011519680.2 Intron XP_011517982.1
XM_011519683.2 Intron XP_011517985.1
XM_011519684.2 Intron XP_011517986.1
XM_011519685.1 Intron XP_011517987.1
XM_011519686.2 Intron XP_011517988.1
XM_011519687.1 Intron XP_011517989.1
XM_011519688.2 Intron XP_011517990.1
XM_017016638.1 Intron XP_016872127.1
XM_017016639.1 Intron XP_016872128.1
XM_017016640.1 Intron XP_016872129.1
XM_017016641.1 Intron XP_016872130.1
XM_017016642.1 Intron XP_016872131.1
XM_017016643.1 Intron XP_016872132.1
XM_017016644.1 Intron XP_016872133.1
XM_017016645.1 Intron XP_016872134.1
XM_017016646.1 Intron XP_016872135.1
XM_017016647.1 Intron XP_016872136.1
XM_017016648.1 Intron XP_016872137.1
XM_017016649.1 Intron XP_016872138.1
XM_017016650.1 Intron XP_016872139.1
XM_017016651.1 Intron XP_016872140.1
XM_017016652.1 Intron XP_016872141.1
XM_017016653.1 Intron XP_016872142.1
Gene
LOC101929431
Gene Name
uncharacterized LOC101929431
There are no transcripts associated with this gene.

View Full Product Details